Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Methanol ≥99.5%, GPR RECTAPUR®
 
undefined
 
 
 
Methanol ≥99.5%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99.5 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.0002 meq/g
  • Boiling point:
    64 - 65 °C
  • Colouration:
    Max. 10 APHA
  • Acetone:
    Max. 10 ppm
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 0.2 %

 

Frequently Bought Together