


Methanol, EMPARTA® ACS analytical reagent, Supelco®
: Merck
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:Ambient
:200-659-6
:1230
:3 (6.1),II
:M-Clarity™ quality level = MQ200
:11 °C (closed cup)
:01-2119433307-44
:GHS02
:GHS06
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Solubility:(20 °C) soluble
- Melting Point:-98 °C
- Molar Mass:32.04 g/mol
- Vapor Pressure:128 hPa (20 °C)
- Flash Point:10 °C
- Refractive Index:1.33
- Explosion Limit:5.5 - 44 %(V)
- Density:0.792 g/cm3 (20 °C)
- pH:64.5 °C (1013 hPa)
- Ignition Point:455 °C DIN 51794
- Evaporation Number:6.3
Frequently Bought Together