


Methanol, BAKER ANALYZED®, Isocratic Grade for HPLC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay:min. 99.8 %
- Color (APHA):max. 10
- Residue after Evaporation (in ppm):max. 5
- Titrable Acid (meq/g):max. 0.0003
- Titrable Base (meq/g):max. 0.0002
- Water (H2O):max. 0.05 %
- at 225 nm:max. 0.30
- at 240 nm:max. 0.10
- at 265 nm:max. 0.01
Specification Test Results
Related Information
Frequently Bought Together