Order Entry
Switzerland
ContactUsLinkComponent
Methanol, BAKER ANALYZED®, gradient grade for HPLC, UHPLC, J.T.Baker®
Flammable
Toxic
Health
Methanol, BAKER ANALYZED®, gradient grade for HPLC, UHPLC, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol, BAKER ANALYZED®, gradient grade for HPLC, UHPLC, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.8 %
  • Acetone:
    max. 0.001 %
  • Residue after Evaporation (in ppm):
    max. 2
  • Titrable Acid (meq/g):
    max. 0.0003
  • Titrable Base (meq/g):
    max. 0.0001
  • Water (H2O):
    max. 0.02 %
  • Measured at Emission Maximum for Solvent Impurities:
    max. 1.0
  • Measured at 450 nm:
    max. 0.3
  • at 235 nm:
    max. 0.005
  • at 254 nm:
    max. 0.001
  • at 225 nm:
    max. 0.20
  • at 280 nm:
    max. 0.01
  • at 350 nm:
    max. 0.01
  • UV Cut-off, nm:
    max. 206

Specification Test Results

Frequently Bought Together