Order Entry
Switzerland
ContactUsLinkComponent
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal®
Toxic
Health
Flammable
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal®
Certificates
undefined
Toxic
Health
Flammable
Methanol, extra dry over Molecular Sieve 99.8%, AcroSeal®
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.8 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g

Specification Test Results

Frequently Bought Together