Ethanol absolute
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
1 Options Available Below
Product Details & Documents
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Glass bottle1 LDiscontinued-
- Glass bottle1 LDiscontinued-
- Glass bottle2,5 LDiscontinued-
- Glass bottle2,5 LDiscontinued-
- Specifications:MarketsourceMarketsource
- 25 LDiscontinued-



