Orders LinkContactUsLinkComponent
Compact burettes, SILBERBRAND
 :   Brand
Compact burettes, SILBERBRAND
 :  Brand
Compact burettes, SILBERBRAND
Compact burettes, SILBERBRAND
Soda-lime glass AR-GLAS®.
  • Calibrated to deliver (TD, Ex)
  • Black graduations
  • Discharge tube with outer diameter 8 mm
Error limits according to class B.
Product Family Options
  • Capacity
    Division
    Length
    Tolerance
  • 25 ml
    0,1 ml
    400 mm
    0,08 ± ml
    1 * 2 ST
    1 * 2 ST - 93.30 SFr.
    93.30 SFr.
    1 * 2 ST
       93.30 SFr.
  • 50 ml
    0,1 ml
    620 mm
    0,10 ± ml
    1 * 2 ST
    1 * 2 ST - 93.30 SFr.
    93.30 SFr.
    1 * 2 ST
       93.30 SFr.
Accessories
  •  
    Description
  • BRND11800
    Stopcock, PTFE with detachable PP tip
    1 * 1 ST - SFr. 37.60
  •  
    Description
  • BRND11600
    Spare burette tip, PP with screw connector
    1 * 1 ST - SFr. 3.00