Order Entry
Switzerland
ContactUsLinkComponent
2-Propanol, anhydrous (max. 0.003% H₂O) ≥99.8%
Flammable
Irritant
2-Propanol, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
2-Propanol, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Evaporation residue:
    Max. 0.0005 %
  • Water:
    Max. 0.0030 %

Specification Test Results

Related Information

Frequently Bought Together