Order Entry
Switzerland
ContactUsLinkComponent
 
Flammable
Irritant
2-Propanol, Prepsolv® for preparative chromatography, Supelco®
Catalog # 1.13350.2500
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
2-Propanol, Prepsolv® for preparative chromatography, Supelco®
Catalog # 1.13350.2500
Supplier:  Merck
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,
 :M-Clarity™ quality level = MQ200
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674