ContactUsLinkComponent
2-Propanol, extra dry over Molecular Sieve 99.5%, AcroSeal®
Irritant
Flammable
2-Propanol, extra dry over Molecular Sieve 99.5%, AcroSeal®
Certificates
undefined
Irritant
Flammable
2-Propanol, extra dry over Molecular Sieve 99.5%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.5 %