Order Entry
Switzerland
ContactUsLinkComponent
2-Propanol, extra dry 99.8%, AcroSeal®
Irritant
Flammable
2-Propanol, extra dry 99.8%, AcroSeal®
Certificates
undefined
Irritant
Flammable
2-Propanol, extra dry 99.8%, AcroSeal®
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.75 %
  • Residue after evaporation:
    ≤0.0005 %

Specification Test Results

Frequently Bought Together