Order Entry
Switzerland
ContactUsLinkComponent
1-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Flammable
Corrosive
Irritant
1-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83635.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Irritant
1-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83635.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.80 %
  • Acidity:
    Max. 0.0005 %
  • Alkalinity:
    Max. 0.0002 %
  • Evaporation residue (100°C):
    Max. 0.0010 %
  • Water:
    Max. 0.0500 %
  • Transmittance (230 nm):
    Min. 50.0 %
  • Transmittance (240 nm):
    Min. 80.0 %
  • Transmittance (270 nm):
    98.0 %
  • Conforms to BDH 15303:
    Passes test

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0.8053 g/cm³ (20 °C. DIN 51757)
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :GHS02
 :GHS05
 :GHS07
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941