Order Entry
Switzerland
Orders LinkContactUsLinkComponent
1-Propanol ≥99.5% ACS

1-Propanol ≥99.5% ACS

Flammable
Corrosive
Irritant
1-Propanol ≥99.5% ACS
Certificates
1-Propanol ≥99.5% ACS
1-Propanol ≥99.5% ACS

2 Options Available Below

Product Details & Documents

Purity: 99.5+% CAS No.: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.10 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.804 Refractive Index: 1.3850 Odour: Alcohol-like Melting Point: -127° Boiling Point: 97° Beilstein Reference: 1098242 Merck Reference: 14,7842 UN No.: 1274 MDL No.: MFCD00002941

Synonyms:  Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
  • CAS Number:71-23-8
  • Formula:H₃CCH₂CH₂OH
  • MDL Number:MFCD00002941
  • Molecular Weight:60.1 g/mol
  • Merck Index:14,7842
  • Boiling Pt:97 °C
  • Melting Pt:–127 °C
  • Density:0.804
  • Flash Pt:15 °C (59 °F)
  • Storage Temperature:Ambient
  • EINECS:200-746-9
  • UN:1274
  • ADR:3,II
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.