Order Entry
Switzerland
Orders LinkContactUsLinkComponent
1-Octanol

1-Octanol

Irritant
Marketsource
Supplier:   Roth Carl
1-Octanol
Supplier:  Roth Carl
Marketsource
Certificates
1-Octanol

2 Options Available Below

Product Details & Documents
Synonyms:  n-Octanol
  • CAS Number:111-87-5
  • Formula:H₃C(CH₂)₆CH₂OH
  • MDL Number:MFCD00002988
  • Molecular Weight:130.23 g/mol
  • Merck Index:13,06784
  • Boiling Pt:195 °C (1013 hPa)
  • Melting Pt:–16 °C
  • Density:0.8246 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-917-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 27.00
    SFr. 27.00
    1 * 100 mL
       SFr. 27.00

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 56.30
    SFr. 56.30
    1 * 500 mL
       SFr. 56.30

    Specifications:
    Marketsource
    Marketsource