ContactUsLinkComponent
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
  1.07026.2500
 :  Merck
 :  
undefined
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
  1.07026.2500
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
 :
 :74.12 g/mol
 :12,03852
 :35 °C (1013 hPa)
 :–123 °C
 :0.7135 g/cm³ (20 °C)
 :–40 °C
 :Ambient
 :200-467-2
 :1155
 :3,I
 :M-Clarity™ quality level = MQ200
 :01-2119535785-29
 :GHS02
 :GHS07
 :60-29-7
 :(CH₃CH₂)₂O
 :MFCD00011646