Diethyl ether 99+% stabilised for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:34.6 °C (760 mmHg)
Melting Pt:–116 °C
Density:0.714 g/cm³
Flash Pt:–45 °C
Storage Temperature:Ambient
UN:1155
ADR:3,I
:Stabilised with ethanol
EINECS:200-467-2
REACH:01-2119535785-29
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear liquid
- GC:≥99.0 % (Excluding stabilizer)
- UV:at 220 nm A: ≤1
- Color scale:≤10 APHA
- Stabilizer:Approx. 1 % w/w Ethanol
- Peroxides:≤5 ppm (as H2O2)
Specification Test Results
Frequently Bought Together