

Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:34.6 °C (760 mmHg)
Melting Pt:–116 °C
Density:0.714 g/cm³
Flash Pt:–45 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
REACH:01-2119535785-29
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Stabilizer:1.5 to 10 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Filt.over 0.2 micron PFTE:Filtered
- Peroxides:≤0.002 % (as H2O2)
Specification Test Results
Related Information
Frequently Bought Together