


Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
65897-50ML
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Identity (IR):complying
- Assay (GC):Min. 99.5 %
- Refractive index (n 20/D):1.400 - 1.402
- Appearance:complying
- Non-volatile matter:Max. 0.005 %
- Water (Karl Fischer):Max. 0.5 %
- Aluminium (Al):Max. 0.2 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.02 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- UV- cutoff:250.0 - 280.0 nm
- Vapor Density:3.5 (vs air)
:
:101.19 g/mol
:13,09740
:90 °C
:-115 °C
:–115 °C
:0.73 g/cm3 (20 °C)
:-11 °C
:–11 °C
:231-913-4
:1296
:3,II
:GHS02
:GHS05
:GHS07
:Ambient
:121-44-8
:(C₂H₅)₃N
:MFCD00009051