


Triethylamine ≥99.5%, BioSyn™ for HPLC, Fluka™
Catalog # HONC17924-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (GC):Min. 99.5 %
- Suitability test (HPLC):complying
- Vapor Pressure:51.75 mmHg ( 20 °C)
- Vapor Density:3.5 (vs air)
- Autoignition Temperature:593 °F
- Explosion Limit:8 %
Specification Test Results
About this item
:
:101.19 g/mol
Merck Index:12,09799
Boiling Pt:90 °C
Melting Pt:-115 °C
Melting Pt:–115 °C
Density:0.73 g/cm³ (20 °C)
Flash Pt:-11 °C
Flash Pt:–11 °C
EINECS:204-469-4
UN:1296
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051