Order Entry
Switzerland
ContactUsLinkComponent
Triethylamine ≥99.5%, BAKER BIO-ANALYZED®, J.T.Baker®
Flammable
Corrosive
Irritant
Triethylamine ≥99.5%, BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # 9111-07
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Triethylamine ≥99.5%, BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # 9111-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay ((CH₃CH₂)₃N):
    ≥ 99.0 %
  • Color (APHA):
    ≤ 15
  • Identification (by IR):
    Passes Test
  • Water (H₂O):
    ≤ 1 %

Specification Test Results

About this item

 :
 :101.19 g/mol
Merck Index:13,09740
Boiling Pt:90 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:204-469-4
UN:1296
ADR:3,II
 :Management of Change (MOC) category = N
 :GHS02
 :GHS05
 :GHS07
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051