Organic reference standard, Trichlorofluoromethane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA13798500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-69-4
- Description:Trichlorofluoromethane 100 µg/ml in methanol
- Formula:CCl₃F
- Description:Trichlorofluoromethane