Order Entry
Switzerland
ContactUsLinkComponent
Chloroform, dehydrated (max. 0.01% H₂O) 99.0-99.6% stabilised, GPR RECTAPUR® for synthesis
Health
Toxic
Chloroform, dehydrated (max. 0.01% H₂O) 99.0-99.6% stabilised, GPR RECTAPUR® for synthesis
Catalog # 22710.321
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Toxic
Chloroform, dehydrated (max. 0.01% H₂O) 99.0-99.6% stabilised, GPR RECTAPUR® for synthesis
Catalog # 22710.321
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    99.0 - 99.6 %
  • Density (20/4):
    1.473 - 1.489
  • Ethanol:
    0.1 - 1.0 %
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 100 ppm

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with ethanol 0.6 %
REACH:01-2119486657-20
 :GHS08
 :GHS06
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826