

Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:M-Clarity™ quality level = MQ300
EINECS:200-663-8
REACH:05-2114614906-45
:GHS06
:GHS08
Specification Test Results
- Solubility:8 g/l (20 °C)
- Melting Point:-63 °C
- Molar Mass:119.38 g/mol
- Boiling Point:61 °C (1013 hPa)
- Vapor Pressure:211 hPa (20 °C)
- Refractive Index:1.45
- Density:1.48 g/cm3 (20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.02445.0250_TDS_EN MERC1.02445.1000_TDS_EN MERC1.02445.2500_TDS_EN MERC1.02445.4000_TDS_EN ProductPage_102445
Product specification
Merck Aquastar® Reagents and Standards for Precise Karl Fischer Titration Results Merck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure
Product Literature
msds_102445
SDS
Frequently Bought Together