

Chloroform 99.0-99.4% (by GC) stabilised, EMPROVE® ESSENTIAL, SAFC®
:67-66-3
:
:CHCl₃
:MFCD00000826
:119.38 g/mol
:13,02160
:61.7 °C (1013 hPa)
:–63 °C
:1.4832 g/cm³ (20 °C)
:Ambient
:1888
:6.1,III
:Stabilised with ethanol 1,0 - 2,0 % (v/v)
:200-663-8
:05-2114614906-45
:GHS06
:GHS08
- Purity (GC):99.0 - 99.4 % (w/w)
- Identity (IR):conforms
- Acidity or alkalinity:conforms
- Density (d 20 °C/ 4 °C):1.474 - 1.479
- Density (d 20 °C/20 °C):1.479 - 1.482
- Boiling range (60-62°C):≥ 95 % (v/v)
- Chloride (Cl):conforms
- Free chlorine (as Cl):conforms
- Ethanol (GC):1.0 - 2.0 % (v/v)
- Related substances (GC):≤ 1.0 % (v/v)
- Bromochloromethane (GC):≤ 0.01 %
- 1,2-Dichloroethene (GC):≤ 0.05 %
- Dichloromethane (GC):≤ 0.01 %
- Carbon tetrachloride (GC):≤ 0.01 %
- 1,1,2-Trichloroethene (GC):≤ 0.01 %
- Other residual solvents (ICH Q3C):excluded by manufacturing process
- Aldehydes:conforms
- Foreign chlorine compounds:conforms
- Matter discolored by H₂SO₄:≤ 10 Hazen
- Evaporation residue:≤ 0.0010 %
- Water:≤ 0.10 %
Frequently Bought Together