Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Chloroform 99-99.4% (by GC) stabilised, meets analytical specification of DAB9, BP, Riedel-de Haën™
 
undefined
 
 
Chloroform 99-99.4% (by GC) stabilised, meets analytical specification of DAB9, BP, Riedel-de Haën™
 

 

  • Appearance of the substance:
    complying
  • Assay (GC):
    99 - 99.4 %
  • Boiling range:
    60 - 62 °C
  • Density (D 20/20):
    1.479 - 1.484
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Acid. or alk. react. impurities:
    complying
  • Reaction against H₂SO₄:
    complying
  • Chloride (Cl):
    Max. 0.0001 %
  • Free chlorine:
    complying
  • Foreign chlorine compounds:
    complying
  • Aldehydes:
    complying
  • Ethanol (GC):
    0.6 - 1.0 Gew.%
  • Related substances (GC):
    complying

 

 

Frequently Bought Together