Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Dichloromethane 99.9% stabilised for peptide synthesis
 
undefined
 
 
Dichloromethane 99.9% stabilised for peptide synthesis

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.005 %
  • GC:
    ≥99.9 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    0.003 to 0.006 % (Amylene)
  • Residue after evaporation:
    ≤0.0005 %
  • Free amine:
    ≤0.001 %

 

 

Frequently Bought Together