

Dichloromethane stabilised for residue analysis, for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–97 °C
Density:1.325 g/cm³
UN:1593
ADR:6.1,III
:Stabilised with amylene
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 % (K.F.)
- GC:(ECD) Passes test, (FID) Passes test
- Stabilizer:Approx. 50 ppm amylene
- Residue after evaporation:≤2 ppm
Specification Test Results
Frequently Bought Together