Order Entry
Switzerland
ContactUsLinkComponent
 
Flammable
1-Chlorobutane, LiChrosolv® for HPLC, Supelco®
Catalog # 1.01692.1000
Supplier:  Merck
CAS Number:  
undefined
Flammable
1-Chlorobutane, LiChrosolv® for HPLC, Supelco®
Catalog # 1.01692.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :92.57 g/mol
Merck Index:13,01559
Boiling Pt:78.4 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.8862 g/cm³ (20 °C)
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:203-696-6
UN:1127
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
CAS Number:109-69-3
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009