
1-Chlorobutane ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-69-3
:
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009
:92.57 g/mol
Merck Index:14,1560
Boiling Pt:77…78 °C
Melting Pt:–123 °C
Density:0.886
Flash Pt:–12 °C (10 °F)
Storage Temperature:Ambient
EINECS:203-696-6
UN:1127
ADR:3,II
:GHS02
Related Information
Frequently Bought Together