Ammonium acetate, Supelco®
Supplier: Merck
Jump to:
About this item
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:13,00495
- Melting Pt:114 °C
- Density:1.17 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:136 °C
- Storage Temperature:Ambient
- EINECS:211-162-9
2 Options Available Below
Product Details & Documents
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:13,00495
- Melting Pt:114 °C
- Density:1.17 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:136 °C
- Storage Temperature:Ambient
- EINECS:211-162-9



