
Ammonium acetate ≥98.0% for soil analysis
21199.361
:
- Pack type:Plastic bucket
- Pk:5 kg
- Seq:4114
- Assay:Min. 98.0 %
- pH (20°C; 5 %):5.0 - 7.4
- Heavy metals (as Pb):Max. 2 ppm
- Ignition residue (SO4):Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- NO3 (Nitrate):Max. 10 ppm
- SO4 (Sulphate):Max. 10 ppm
- Ca (Calcium):Max. 5 ppm
- Cu (Copper):Max. 0.5 ppm
- Fe (Iron):Max. 2 ppm
- K (Potassium):Max. 1 ppm
- Mg (Magnesium):Max. 1 ppm
- Na (Sodium):Max. 1 ppm
- Pb (Lead):Max. 0.5 ppm
- Zn (Zinc):Max. 0.5 ppm
:
:77.08 g/mol
:13,00495
:114 °C
:1.17 g/cm³ (20 °C)
:136 °C
:211-162-9
:GHS07
:Ambient
:631-61-8
:H₃CCOONH₄
:MFCD00013066