Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ammonium acetate

Ammonium acetate

Glass bottle for solids, 250 g
Catalog # MSPPA/3446/50
Irritant
Marketsource
Ammonium acetate
Catalog # MSPPA/3446/50
Marketsource
CAS Number:  631-61-8
Ammonium acetate
Ammonium acetate

Glass bottle for solids, 250 g

CAS Number:  631-61-8
Price
Inquire For Price
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Melting Pt:114 °C
  • Density:1.17 g/cm³ (20 °C)
  • Flash Pt:136 °C
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.