Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tridecanoic acid

Tridecanoic acid

Glass bottle for solids, 100 mg
Catalog # MAYA1161
Irritant
Supplier:   MATREYA
Tridecanoic acid
Catalog # MAYA1161
Supplier:  MATREYA
CAS Number:  638-53-9
Tridecanoic acid

Glass bottle for solids, 100 mg

CAS Number:  638-53-9
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00002741
  • Molecular Weight:214.35 g/mol
  • Boiling Pt:308 °C (1013 hPa)
  • Melting Pt:41.5 °C
  • Density:1.025 g/cm³ (20 °C)
  • Storage Temperature:Freezer
  • EINECS:211-341-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications