Tridecanoic acid 97%
: n-Tridecanoic acid, Tridecylic acid
- :638-53-9
- :C₁₃H₂₆O₂
- :MFCD00002741
- :214.35 g/mol
- :236 °C (100 mmHg)
- :40…45 °C
- :1,025 g/cm³ (20 °C)
- :113 °C
- :Ambient
- :211-341-1
: n-Tridecanoic acid, Tridecylic acid
- :638-53-9
- :C₁₃H₂₆O₂
- :MFCD00002741
- :214.35 g/mol
- :236 °C (100 mmHg)
- :40…45 °C
- :1,025 g/cm³ (20 °C)
- :113 °C
- :Ambient
- :211-341-1
- Appearance:White to light yellow Crystalline powder or flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:40 to 45 °C
- Titration with NaOH:≥96.0 %
Product Family Options
- Pack typePk



