Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Propionic acid, Supelco®

Corrosive
Irritant
Supplier:   Merck
Propionic acid, Supelco®
Supplier:  Merck
Certificates
Propionic acid, Supelco®
Propionic acid, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:0.99 g/cm³ (20 °C)
  • Flash Pt:49 °C
  • UN:3463
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • REACH:01-2119486971-24

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - SFr. 54.50
    SFr. 54.50
    1 * 1 mL
       SFr. 54.50

  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - SFr. 140.00
    SFr. 140.00
    1 * 5 mL
       SFr. 140.00