Propionic acid 99+%, extra pure
: Propanoic acid
- :79-09-4
- :C₃H₆O₂
- :MFCD00002756
- :74.08 g/mol
- :13,07917
- :141 °C (1013 hPa)
- :–22 °C
- :0.99 g/cm³
- :51 °C
- :Ambient
- :201-176-3
- :3463
- :8,II
- :01-2119486971-24
: Propanoic acid
- :79-09-4
- :C₃H₆O₂
- :MFCD00002756
- :74.08 g/mol
- :13,07917
- :141 °C (1013 hPa)
- :–22 °C
- :0.99 g/cm³
- :51 °C
- :Ambient
- :201-176-3
- :3463
- :8,II
- :01-2119486971-24
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3850 to 1.3870 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥99.0 %
- Color scale:≤10 APHA
Product Family Options
- Pack typePk



