Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pentadecanoic acid

Pentadecanoic acid

Glass bottle for solids, 1 g
Catalog # MAYA1012
Supplier:   MATREYA
Pentadecanoic acid
Catalog # MAYA1012
Supplier:  MATREYA
CAS Number:  1002-84-2
Pentadecanoic acid

Glass bottle for solids, 1 g

CAS Number:  1002-84-2
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Pentadecylic acidn-Pentadecanoic acid
  • CAS Number:1002-84-2
  • Formula:C₁₅H₃₀O₂
  • MDL Number:MFCD00002745
  • Molecular Weight:242.4 g/mol
  • Boiling Pt:339 °C (1013 hPa)
  • Melting Pt:51…53 °C
  • Density:0.8496 g/cm³ (70 °C)
  • Storage Temperature:Ambient
  • EINECS:213-693-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.