Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pentadecanoic acid 99%

Pentadecanoic acid 99%

Pentadecanoic acid 99%
Certificates
Pentadecanoic acid 99%

2 Options Available Below

Product Details & Documents
Synonyms:  Pentadecylic acidn-Pentadecanoic acid
  • CAS Number:1002-84-2
  • Formula:C₁₅H₃₀O₂
  • MDL Number:MFCD00002745
  • Molecular Weight:242.4 g/mol
  • Boiling Pt:339 °C (1013 hPa)
  • Melting Pt:51…53 °C
  • Density:0,8496 g/cm³ (70 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:213-693-1

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals or flakes and/or chunks
  • Infrared spectrum:
    Conforms
  • GC:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %
  • Specific optical rotation:
    -63° to -67° (20°C, 589 nm) (c=4, methanol)
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.