
Stearic acid ≥97%, GPR RECTAPUR®
Catalog # 30267HG
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Assay:Min. 97 %
- IR Spectrum:Passes test
- Melting point:68 - 70 °C
Specification Test Results
About this item
:
:284.48 g/mol
Merck Index:13,08882
Boiling Pt:361 °C (1013 hPa)
Melting Pt:67 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:196 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:200-313-4
REACH:01-2119543894-28
:GHS07
CAS Number:57-11-4
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00002752