Nonadecanoic acid
Glass bottle for solids, 100 mg
MAYA1028
: MATREYA
: Nonadecylic acid
- :646-30-0
- :C₁₉H₃₈O₂
- :MFCD00002754
- :298.51 g/mol
- :297 °C (1013 hPa)
- :68…70 °C
- :0,8468 g/cm³ (70 °C)
- :Ambient
- :211-472-4
- Pack type:Glass bottle for solids
- Pk:100 mg