Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Nonadecanoic acid 98+%

Nonadecanoic acid 98+%

Irritant
Nonadecanoic acid 98+%
Certificates
Nonadecanoic acid 98+%

2 Options Available Below

Product Details & Documents
Synonyms:  Nonadecylic acid
  • CAS Number:646-30-0
  • Formula:C₁₉H₃₈O₂
  • MDL Number:MFCD00002754
  • Molecular Weight:298.51 g/mol
  • Boiling Pt:297 °C (100 mmHg)
  • Melting Pt:68…70 °C
  • Density:0,8468 g/cm³ (70 °C)
  • Storage Temperature:Ambient
  • EINECS:211-472-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Glistening powder or flakes
  • Infrared spectrum:
    Conforms
  • GC:
    ≥98.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 61.60
    SFr. 61.60
    1 * 1 g
       SFr. 61.60

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 153.00
    SFr. 153.00
    1 * 5 g
       SFr. 153.00