Formic acid 98%
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:1779
3 Options Available Below
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:1779
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



