

Acetic acid ≥96%, EMPROVE® ESSENTIAL, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 (3),II
:GHS02
:GHS05
Related Information
Frequently Bought Together