

Acetic acid 99.6% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Melting Pt:16…16.5 °C
Density:1.048 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.2 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Titration with NaOH:≥99.5 %
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
Specification Test Results
Frequently Bought Together