Behenic acid
Glass bottle for solids, 500 mg
Catalog # MAYA1035
Supplier: MATREYA
Jump to:
Product Details & Documents
Synonyms: Docosanoic acid
- CAS Number:112-85-6
- Formula:C₂₂H₄₄O₂
- MDL Number:MFCD00002807
- Molecular Weight:340.59 g/mol
- Merck Index:13,01024
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:80 °C
- Density:0.8221 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-010-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



