Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Behenic acid 85%, Technical Grade

Behenic acid 85%, Technical Grade

Behenic acid 85%, Technical Grade
Certificates
Behenic acid 85%, Technical Grade

2 Options Available Below

Product Details & Documents
Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:13,01024
  • Boiling Pt:306 °C (60 mmHg)
  • Melting Pt:80…82 °C
  • Density:0,8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥80 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - SFr. 57.10
    SFr. 57.10
    1 * 250 g
       SFr. 57.10

  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 192.00
    SFr. 192.00
    1 * 1 kg
       SFr. 192.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.