Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Capric acid

Capric acid

Glass bottle for solids, 1 g
Catalog # ROTH3164.1
Irritant
Marketsource
Supplier:   Roth Carl
Capric acid
Catalog # ROTH3164.1
Supplier:  Roth Carl
Marketsource
CAS Number:  334-48-5
Capric acid

Glass bottle for solids, 1 g

CAS Number:  334-48-5
Price
SFr. 38.30
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Decylic acid, Decanoic acid
  • CAS Number:334-48-5
  • Formula:C₁₀H₂₀O₂
  • MDL Number:MFCD00004441
  • Molecular Weight:172.27 g/mol
  • Merck Index:13,01764
  • Boiling Pt:268 °C (1013 hPa)
  • Melting Pt:29…32 °C
  • Density:0.893 g/cm³ (20 °C)
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:206-376-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications