Order Entry
Switzerland
ContactUsLinkComponent
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Flammable
Irritant
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Residue on evaporation:
    Max. 0.0005 %
  • Water:
    Max. 0.0050 %

Specification Test Results

Related Information

Frequently Bought Together