Supelco® Organic Reference Standard, Butyl acetate
: Merck
- GHS signal: GHS02, GHS07
: P210
- CAS No.:123-86-4
- Storage conditions:2...30 °C
- Hazard class description:H226 - H336
- Shipping conditions:STD
- MDL No.:MFCD00009445
- M-Clarity quality level:MQ100
- Melting point:–78 °C (lit.)
- Formula:CH₃COO(CH₂)₃CH₃
- Molecular weight:116,16
- Density:0,88 g/ml at 25 °C (lit.)
- Flash point:27 °C - Closed cup
- Boiling point:124 - 126 °C (lit.)
Frequently Bought Together