

Isopropyl acetate ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
:108-21-4
:
:C₅H₁₀O₂
:MFCD00008877
:102.13 g/mol
:13,05225
:85 °C
:–62 °C
:0.8662 g/cm³ (25 °C)
:2.2 °C
:Ambient
:203-561-1
:1220
:3,II
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- assay (GC):Min. 99 %
- measurement uncertainty:effective %
- identity (NMR):complying
- refractive index (n 25/D):1.38 - 1.39
- water (Karl Fischer):Max. 0.5 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Autoignition Temperature:894 °F
Frequently Bought Together